Learn More
Cayman Chemical DBCO acid, Purity: ≥98%, CAS: 1353016-70-2, 500 mg
SDP
Catalog No. 502942621
Encompass_Preferred
A copper-free click chemistry reagent; has been used in the study of glycoprotein charge effects on the migration and invasion of 9-azido sialic acid-labeled cancer cells in vitro; has also been used in the synthesis of DBCO-modified RNase A to deliver RNase A into AAM-labeled cancer cells in vitro and in vivo to induce cytotoxicity and reduce tumor growth, respectively
Catalog No. 50-294-2621 Supplier Cayman Chemical Supplier No. 41894500 MG
A copper-free click chemistry reagent; has been used in the study of glycoprotein charge effects on the migration and invasion of 9-azido sialic acid-labeled cancer cells in vitro; has also been used in the synthesis of DBCO-modified RNase A to deliver RNase A into AAM-labeled cancer cells in vitro and in vivo to induce cytotoxicity and reduce tumor growth, respectively