Learn More
Cayman Chemical ANTIBODY DBCO acId 50mg
SDP
Catalog No. 502944109
Encompass_Preferred
A copper-free click chemistry reagent; has been used in the study of glycoprotein charge effects on the migration and invasion of 9-azido sialic acid-labeled cancer cells in vitro; has also been used in the synthesis of DBCO-modified RNase A to deliver RNase A into AAM-labeled cancer cells in vitro and in vivo to induce cytotoxicity and reduce tumor growth, respectively
Catalog No. 50-294-4109 Supplier Cayman Chemical Supplier No. 4189450 MG
A copper-free click chemistry reagent; has been used in the study of glycoprotein charge effects on the migration and invasion of 9-azido sialic acid-labeled cancer cells in vitro; has also been used in the synthesis of DBCO-modified RNase A to deliver RNase A into AAM-labeled cancer cells in vitro and in vivo to induce cytotoxicity and reduce tumor growth, respectively