Learn More
Cayman Chemical DBCO-amine, Purity: ≥98%, CAS: 1255942-06-3, 250 mg
SDP
Catalog No. 502943692
Encompass_Preferred
A copper-free click chemistry reagent; has been used in the synthesis of clickable fluorescent probes
Catalog No. 50-294-3692 Supplier Cayman Chemical Supplier No. 41895250 MG
A copper-free click chemistry reagent; has been used in the synthesis of clickable fluorescent probes