Learn More
Cayman Chemical ANTIBODY DBCO-amIn 500mg
SDP
Catalog No. 502943872
Encompass_Preferred
A copper-free click chemistry reagent; has been used in the synthesis of clickable fluorescent probes
Catalog No. 50-294-3872 Supplier Cayman Chemical Supplier No. 41895500 MG
A copper-free click chemistry reagent; has been used in the synthesis of clickable fluorescent probes