Learn More
Cayman Chemical ANTIBODY DBCO-amIn 50mg
SDP
Catalog No. 502943804
Encompass_Preferred
A copper-free click chemistry reagent; has been used in the synthesis of clickable fluorescent probes
Catalog No. 50-294-3804 Supplier Cayman Chemical Supplier No. 4189550 MG
A copper-free click chemistry reagent; has been used in the synthesis of clickable fluorescent probes