Learn More
Cayman Chemical DC-05, Purity: ≥98%, CAS: 890643-16-0, 10 mg
SDP
Catalog No. 502913802
Encompass_Preferred
A non-nucleoside DNMT1 inhibitor (IC50 = 10.3 µM); selective for DNMT1 over DNMT3A and DNMT3B (IC50s = >200 µM), as well as G9a, SUV39H1, MLL1, SET7/9, and PRMT1 (IC50s = >150, >150, >150, >150, and 37.1 µM, respectively); reduces proliferation of HCT116 colon cancer cells from 2.5-10 µM
Catalog No. 50-291-3802 Supplier Cayman Chemical Supplier No. 3658410 MG
A non-nucleoside DNMT1 inhibitor (IC50 = 10.3 µM); selective for DNMT1 over DNMT3A and DNMT3B (IC50s = >200 µM), as well as G9a, SUV39H1, MLL1, SET7/9, and PRMT1 (IC50s = >150, >150, >150, >150, and 37.1 µM, respectively); reduces proliferation of HCT116 colon cancer cells from 2.5-10 µM