Learn More
Cayman Chemical DCP-Rho1, Purity: ≥98%, CAS: 1001575-98-9, 1 mg
SDP
Catalog No. 502955938
Encompass_Preferred
A fluorescent probe for sulfenic acid-containing proteins; ex/em = 560/581 nm, respectively; has been used to visualize protein oxidation sites in situ
Catalog No. 50-295-5938 Supplier Cayman Chemical Supplier No. 131941 MG
A fluorescent probe for sulfenic acid-containing proteins; ex/em = 560/581 nm, respectively; has been used to visualize protein oxidation sites in situ