Learn More
Cayman Chemical Defactinib, Purity: ≥95%, CAS: 1073154-85-4, 5 mg
SDP
Catalog No. 502980091
Encompass_Preferred
A dose-dependent inhibitor of FAK, with maximal inhibition of FAK autophosphorylation in cells achieved at 10 µM; restores the chemosensitivity of taxane-resistant cells to paclitaxel; orally bioavailable, inhibiting FAK and augmenting paclitaxel action against ovarian cancer cell tumors in mice
Catalog No. 50-298-0091 Supplier Cayman Chemical Supplier No. 177375 MG
A dose-dependent inhibitor of FAK, with maximal inhibition of FAK autophosphorylation in cells achieved at 10 µM; restores the chemosensitivity of taxane-resistant cells to paclitaxel; orally bioavailable, inhibiting FAK and augmenting paclitaxel action against ovarian cancer cell tumors in mice