Learn More
Cayman Chemical DFHBI 1T, Purity: ≥98%, CAS: 1539318-36-9, 5 mg
SDP
Catalog No. 502946828
Encompass_Preferred
A fluorescent probe for RNA; ex/em = 447/501 nm, respectively; fluoresces upon binding to RNA aptamers; has been used to image RNA in live cells
Catalog No. 50-294-6828 Supplier Cayman Chemical Supplier No. 335175 MG
A fluorescent probe for RNA; ex/em = 447/501 nm, respectively; fluoresces upon binding to RNA aptamers; has been used to image RNA in live cells