Learn More
Cayman Chemical DG-051, Purity: ≥98%, CAS: 929915-58-2, 1 mg
SDP
Catalog No. 502986000
Encompass_Preferred
A potent LTA4H inhibitor (Kd = 25 nM; IC50 = 47 nM); also inhibits the aminopeptidase activity of LTA4H (IC50s = 72 -150 nM); does not show inhibitory activity against a panel of more than 50 additional aminopeptidases, ion channels, or HERG
Catalog No. 50-298-6000 Supplier Cayman Chemical Supplier No. 216881 MG
A potent LTA4H inhibitor (Kd = 25 nM; IC50 = 47 nM); also inhibits the aminopeptidase activity of LTA4H (IC50s = 72 -150 nM); does not show inhibitory activity against a panel of more than 50 additional aminopeptidases, ion channels, or HERG