Learn More
Cayman Chemical ANTIBODY DIapocynIn 5mg
SDP
Catalog No. 502955949
Encompass_Preferred
An NADPH oxidase inhibitor with anti-inflammatory and antioxidant activities; inhibits NADPH oxidase complex assembly and activation, gp91phox mRNA expression, and production of the pro-inflammatory cytokines TNF-α and IL-10 in PMBCs; inhibits ROS production, iPLA2 function, and reduces Ca2+ influx through SOCs and SACs in dystrophic myotubes; reverses motor coordination deficits in the LRRK2R1441G mouse model of EArly Parkinson's disease
Catalog No. 50-295-5949 Supplier Cayman Chemical Supplier No. 140525 MG
An NADPH oxidase inhibitor with anti-inflammatory and antioxidant activities; inhibits NADPH oxidase complex assembly and activation, gp91phox mRNA expression, and production of the pro-inflammatory cytokines TNF-α and IL-10 in PMBCs; inhibits ROS production, iPLA2 function, and reduces Ca2+ influx through SOCs and SACs in dystrophic myotubes; reverses motor coordination deficits in the LRRK2R1441G mouse model of EArly Parkinson's disease