Learn More
Cayman Chemical Dibucaine, Purity: ≥98%, CAS: 85-79-0, 5 mg
SDP
Catalog No. 502964012
Encompass_Preferred
An anesthetic that acts as a sodium channel blocker
Catalog No. 50-296-4012 Supplier Cayman Chemical Supplier No. 222375 MG
An anesthetic that acts as a sodium channel blocker