Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antibody Diluent

Catalog No. 5075038
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-750-38 250 mL
50-750-39 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-750-38 Supplier LIFE TECHNOLOGIES Supplier No. 003118
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Ready-to-use diluent with BSA (for stabilizing the antibody) and preservative.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Diluent
Quantity 250 mL
Form Liquid
Content And Storage Store in refrigerator at 2°C to 8°C
  • Shelf life: 18 months from date of manufacture
  • Shipping Condition Wet Ice
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.