Learn More
Electron Microscopy Sciences Universal Antibody Dilution Buffer  500 ML
SDP
Catalog No. 5033571
Encompass_Preferred
This ready-to-use Universal Antibody Dilution Buffer is designed for dilution of antibodies in all immunoassays, including immunofluorescence, IHC, ELISA, WB. This buffer does not contain any mammalian proteins, phosphate, sodium azide, or mercury preservatives and can be used for dilution of all antibodies, including peroxidase, and antibodies to phophoproteins. However, it is not suitable for dilution of antibodies to S100 proteins.
Ready to use buffer supplied. Dilute antibodies, primary or secondary as recommended in the protocol by the manufacturer of these products. Diluted antibodies can be stored at 2–8 degree C for several weeks, depending upon the stability of antibodies.
Catalog No. 50-335-71 Supplier Electron Microscopy Sciences Supplier No. 2588505
This ready-to-use Universal Antibody Dilution Buffer is designed for dilution of antibodies in all immunoassays, including immunofluorescence, IHC, ELISA, WB. This buffer does not contain any mammalian proteins, phosphate, sodium azide, or mercury preservatives and can be used for dilution of all antibodies, including peroxidase, and antibodies to phophoproteins. However, it is not suitable for dilution of antibodies to S100 proteins.
Ready to use buffer supplied. Dilute antibodies, primary or secondary as recommended in the protocol by the manufacturer of these products. Diluted antibodies can be stored at 2–8 degree C for several weeks, depending upon the stability of antibodies.