Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Antibody Dilution Buffer, free BSA 100 ML
SDP

Catalog No. 5033568
Encompass_Preferred

This ready-to-use Antibody Dilution Buffer is designed for the dilution of antibodies in all immunoassays, including immunofluorescence, IHC, ELISA, WB. This buffer contains immunoglobulin free BSA and does not contain phosphate, sodium azide, or mercury preservatives. It can be used for the dilution of all antibodies, including peroxidase, and antibodies to phophoproteins. However, it is not suitable for dilution of antibodies to S100 proteins. This buffer contains green food color.

Application: Dilute antibodies, primary or secondary as recommended in the protocol by the manufacturer of these products.
Storage: 2 – 8 degree C. Diluted antibodies can be stored at 2 – 8 degree C for several weeks, depending upon the stability of antibodies.

Catalog No. 50-335-68 Supplier Electron Microscopy Sciences Supplier No. 2588601
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This ready-to-use Antibody Dilution Buffer is designed for the dilution of antibodies in all immunoassays, including immunofluorescence, IHC, ELISA, WB. This buffer contains immunoglobulin free BSA and does not contain phosphate, sodium azide, or mercury preservatives. It can be used for the dilution of all antibodies, including peroxidase, and antibodies to phophoproteins. However, it is not suitable for dilution of antibodies to S100 proteins. This buffer contains green food color.

Application: Dilute antibodies, primary or secondary as recommended in the protocol by the manufacturer of these products.
Storage: 2 – 8 degree C. Diluted antibodies can be stored at 2 – 8 degree C for several weeks, depending upon the stability of antibodies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.