Learn More
Cayman Chemical Dianemycin, Purity: ≥95%, CAS: 35865-33-9, 25 mg
SDP
Catalog No. 502964271
Encompass_Preferred
A polyether ionophore antibiotic; has insecticidal and antibacterial properties; blocks ZIKV entry into U2OS, human microvascular endothelial cells, and placental Jeg3 cells (IC50s = 100, 400, and 970 nM, respectively); blocks ZIKV entry into certain primary cells; effective against West Nile, dengue, Sindbis, and chikungunya viruses in U2OS cells
Catalog No. 50-296-4271 Supplier Cayman Chemical Supplier No. 2167925 MG
A polyether ionophore antibiotic; has insecticidal and antibacterial properties; blocks ZIKV entry into U2OS, human microvascular endothelial cells, and placental Jeg3 cells (IC50s = 100, 400, and 970 nM, respectively); blocks ZIKV entry into certain primary cells; effective against West Nile, dengue, Sindbis, and chikungunya viruses in U2OS cells