Learn More
Cayman Chemical Diprotin A, Purity: ≥98%, CAS: 90614-48-5, 25 mg
SDP
Catalog No. 502941799
Encompass_Preferred
A tripeptide inhibitor of DPP-4 (IC50 = 1.1 μg/ml); inhibits degradation of GLP-1 in culture with insulin-secreting BRIN-BD11 rat pancreatic β-cells at 25 μM; pre-incubation at 5 mg/ml enhances chemotaxis of murine embryonic stem cells towards SDF-1/CXCL12 in vitro; pre-incubation of CD34+ human umbilical cord blood cells with 5 mM prior to injection increases engraftment in NOD/SCID recipient mice
Catalog No. 50-294-1799 Supplier Cayman Chemical Supplier No. 2801425 MG
A tripeptide inhibitor of DPP-4 (IC50 = 1.1 μg/ml); inhibits degradation of GLP-1 in culture with insulin-secreting BRIN-BD11 rat pancreatic β-cells at 25 μM; pre-incubation at 5 mg/ml enhances chemotaxis of murine embryonic stem cells towards SDF-1/CXCL12 in vitro; pre-incubation of CD34+ human umbilical cord blood cells with 5 mM prior to injection increases engraftment in NOD/SCID recipient mice