Learn More
Cayman Chemical Diuron, Purity: ≥98%, CAS: 330-54-1, 100 mg
SDP
Catalog No. 502933315
Encompass_Preferred
An herbicide; decreases total respiration in roots of wheat by 50% and ground respiration by 100% at 25 nM/ml; increases the incidence of urinary bladder urothelial carcinomas in male and female mice by 73 and 27%, respectively at 2,500 pm in the diet; increases ROS in MCF-7 and BeWo cells at 200 µM; reduces viability of BeWo cells at 50 and 200 µM
Catalog No. 50-293-3315 Supplier Cayman Chemical Supplier No. 24040100 MG
An herbicide; decreases total respiration in roots of wheat by 50% and ground respiration by 100% at 25 nM/ml; increases the incidence of urinary bladder urothelial carcinomas in male and female mice by 73 and 27%, respectively at 2,500 pm in the diet; increases ROS in MCF-7 and BeWo cells at 200 µM; reduces viability of BeWo cells at 50 and 200 µM