Learn More
Cayman Chemical DLAC, Purity: ≥97%, CAS: 70803-56-4, 250 mg
SDP
Catalog No. 503015997
Encompass_Preferred
A detergent used to solubilize membrane proteins; synthesized from lactobionic acid; CMC = 1.3 mM
Catalog No. 50-301-5997 Supplier Cayman Chemical Supplier No. 24780250 MG
A detergent used to solubilize membrane proteins; synthesized from lactobionic acid; CMC = 1.3 mM