Learn More
Cayman Chemical DLin-DMA, Purity: ≥95%, CAS: 871258-12-7, 100 mg
SDP
Catalog No. 502909307
Encompass_Preferred
An ionizable amino lipid; has been used in combination with other lipids in the formation of LNPs
Catalog No. 50-290-9307 Supplier Cayman Chemical Supplier No. 36701100 MG
An ionizable amino lipid; has been used in combination with other lipids in the formation of LNPs