Learn More
Cayman Chemical DLin-MeOH, Purity: ≥98%, CAS: 1169768-28-8, 10 mg
SDP
Catalog No. 502942596
Encompass_Preferred
A synthetic intermediate in the synthesis of DLin-MC3-DMA
Catalog No. 50-294-2596 Supplier Cayman Chemical Supplier No. 4143210 MG
A synthetic intermediate in the synthesis of DLin-MC3-DMA