Learn More
Cayman Chemical DM1-SMCC, Purity: ≥98%, CAS: 1228105-51-8, 25 mg
SDP
Catalog No. 502995034
Encompass_Preferred
A reactive drug-linker molecule containing DM1; inhibits the proliferation of HCC1954 and MDA-MB-468 cells (IC50s = 17.2 and 49.9 nM, respectively); inhibits the proliferation of MCF-7 breast cancer cells (IC50 = 11 nM) when linked to B38.1
Catalog No. 50-299-5034 Supplier Cayman Chemical Supplier No. 2392625 MG
A reactive drug-linker molecule containing DM1; inhibits the proliferation of HCC1954 and MDA-MB-468 cells (IC50s = 17.2 and 49.9 nM, respectively); inhibits the proliferation of MCF-7 breast cancer cells (IC50 = 11 nM) when linked to B38.1