Learn More
Cayman Chemical DMOG, Purity: ≥98%, CAS: 89464-63-1, 10 mg
SDP
Catalog No. 502894872
Encompass_Preferred
A cell permeable, competitive inhibitor of HIF-1α prolyl hydroxylase; stabilizes HIF-1α expression at normal oxygen tensions in cultured cells at concentrations between 0.1 and 1 mM
Catalog No. 50-289-4872 Supplier Cayman Chemical Supplier No. 7121010 MG
A cell permeable, competitive inhibitor of HIF-1α prolyl hydroxylase; stabilizes HIF-1α expression at normal oxygen tensions in cultured cells at concentrations between 0.1 and 1 mM