Learn More
Cayman Chemical DMU2105, Purity: ≥98%, CAS: 1031063-36-1, 50 mg
SDP
Catalog No. 502918297
Encompass_Preferred
A CYP1B1 inhibitor (IC50 = 0.01 µM); selective for CYP1B1 over CYP1A1, CYP1A2, CYP3A4, and CYP2D6 (IC50s = 0.742, 1.2, 18, and 10 µM, respectively); enhances cisplatin-induced cell death in HEK293 cells overexpressing CYP1B1
Catalog No. 50-291-8297 Supplier Cayman Chemical Supplier No. 3852250 MG
A CYP1B1 inhibitor (IC50 = 0.01 µM); selective for CYP1B1 over CYP1A1, CYP1A2, CYP3A4, and CYP2D6 (IC50s = 0.742, 1.2, 18, and 10 µM, respectively); enhances cisplatin-induced cell death in HEK293 cells overexpressing CYP1B1