Learn More
Cayman Chemical Danicopan, Purity: ≥98%, CAS: 1903768-17-1, 10 mg
SDP
Catalog No. 502950236
Encompass_Preferred
A Factor D inhibitor (IC50 = 15 nM in a cell-free assay); selective for Factor D over a panel of 12 serine proteases when used at a concentration of 10 µM; inhibits hemolysis of erythrocytes isolated from patients with paroxysmal nocturnal hemoglobinuria (IC50s = 4-27 nM); suppresses APC activity in monkeys at 200 mg/kg, p.o.
Catalog No. 50-295-0236 Supplier Cayman Chemical Supplier No. 3273710 MG
A Factor D inhibitor (IC50 = 15 nM in a cell-free assay); selective for Factor D over a panel of 12 serine proteases when used at a concentration of 10 µM; inhibits hemolysis of erythrocytes isolated from patients with paroxysmal nocturnal hemoglobinuria (IC50s = 4-27 nM); suppresses APC activity in monkeys at 200 mg/kg, p.o.