Learn More
Cayman Chemical Danoprevir, Purity: ≥95%, CAS: 850876-88-9, 5 mg
SDP
Catalog No. 502933237
Encompass_Preferred
An HCV NS3/4A protease inhibitor (IC50 = 1 nM); selective for NS3/4A over a panel of 53 proteases (10 μM); inhibits replication of the HCV genotypes 1a, 1b, 4, 5, and 6 (IC50s = 0.2-0.4 nM) as well as 2b and 3a (IC50s = 1.6 and 3.5 nM, respectively) in vitro; reduces the number of HCV genotype 1b replicons in Huh-7 cells (EC50 = 1.8 nM)
Catalog No. 50-293-3237 Supplier Cayman Chemical Supplier No. 241855 MG
An HCV NS3/4A protease inhibitor (IC50 = 1 nM); selective for NS3/4A over a panel of 53 proteases (10 μM); inhibits replication of the HCV genotypes 1a, 1b, 4, 5, and 6 (IC50s = 0.2-0.4 nM) as well as 2b and 3a (IC50s = 1.6 and 3.5 nM, respectively) in vitro; reduces the number of HCV genotype 1b replicons in Huh-7 cells (EC50 = 1.8 nM)