Learn More
Cayman Chemical DNP-INT, Purity: ≥98%, CAS: 69311-70-2, 10 mg
SDP
Catalog No. 502892735
Encompass_Preferred
An electron transport inhibitor in plants; competitively inhibits plastoquinol oxidation by binding at the Qo site of cytochrome b6f (Kd = 1.4 nM); inhibits electron flow from water to NADP or methylviologen by 50 and 100% when used at concentrations of 0.5 or 5 µM, respectively
Catalog No. 50-289-2735 Supplier Cayman Chemical Supplier No. 900306110 MG
An electron transport inhibitor in plants; competitively inhibits plastoquinol oxidation by binding at the Qo site of cytochrome b6f (Kd = 1.4 nM); inhibits electron flow from water to NADP or methylviologen by 50 and 100% when used at concentrations of 0.5 or 5 µM, respectively