Learn More
Cayman Chemical Dofequidar, Purity: ≥98%, CAS: 129716-58-1, 1 mg
SDP
Catalog No. 502989885
Encompass_Preferred
A quinoline derivative and inhibitor of multidrug resistance; inhibits P-glycoprotein in a photolabeling assay at 100 μM; increases intracellular accumulation of [3H]-vincristine in HL-60R cells endogenously expressing the gene encoding MRP at 10 μM; restores susceptibility to vincristine- or doxorubicin-induced cytotoxicity in vincristine-resistant P388, vincristine-resistant K562, and doxorubicin-resistant K562 cells; increases survival in a vincristine-resistant P388 murine leukemia model compared with untreated controls at 80 mg/kg twice per day when administered in combination with vincristine
Catalog No. 50-298-9885 Supplier Cayman Chemical Supplier No. 309251 MG
A quinoline derivative and inhibitor of multidrug resistance; inhibits P-glycoprotein in a photolabeling assay at 100 μM; increases intracellular accumulation of [3H]-vincristine in HL-60R cells endogenously expressing the gene encoding MRP at 10 μM; restores susceptibility to vincristine- or doxorubicin-induced cytotoxicity in vincristine-resistant P388, vincristine-resistant K562, and doxorubicin-resistant K562 cells; increases survival in a vincristine-resistant P388 murine leukemia model compared with untreated controls at 80 mg/kg twice per day when administered in combination with vincristine