Learn More
Cayman Chemical Dooku1, Purity: ≥98%, CAS: 2253744-54-4, 10 mg
SDP
Catalog No. 502919066
Encompass_Preferred
An antagonist of yoda1-activated Piezo1 (IC50 = 1.3 µM in HEK293 cells expressing the human receptor); selective for yoda1-actived Piezo1 over constituitively active Piezo1, TRPV4, and TRPC4 at 10 µM; reduces the percentage of cells with yoda1-induced extracellular phosphatidylserine exposure in RBCs isolated from patients with sickle cell anemia at 10 µM; inhibits yoda1-induced relaxation of isolated mouse aortic rings at 10 µM
Catalog No. 50-291-9066 Supplier Cayman Chemical Supplier No. 3916110 MG
An antagonist of yoda1-activated Piezo1 (IC50 = 1.3 µM in HEK293 cells expressing the human receptor); selective for yoda1-actived Piezo1 over constituitively active Piezo1, TRPV4, and TRPC4 at 10 µM; reduces the percentage of cells with yoda1-induced extracellular phosphatidylserine exposure in RBCs isolated from patients with sickle cell anemia at 10 µM; inhibits yoda1-induced relaxation of isolated mouse aortic rings at 10 µM