Learn More
Cayman Chemical DSRM-3716, Purity: ≥95%, CAS: 58142-99-7, 5 g
SDP
Catalog No. 502917349
Encompass_Preferred
A SARM1 inhibitor (IC50 = 75 nM); decreases levels of cADPR and increases NAD+ levels in axotomized primary mouse DRG axons from 1.1-30 µM; prevents axonal degeneration in primary wild-type but not Sarm1-/- axotomized DRG axons
Catalog No. 50-291-7349 Supplier Cayman Chemical Supplier No. 395175 G
A SARM1 inhibitor (IC50 = 75 nM); decreases levels of cADPR and increases NAD+ levels in axotomized primary mouse DRG axons from 1.1-30 µM; prevents axonal degeneration in primary wild-type but not Sarm1-/- axotomized DRG axons