Learn More
Cayman Chemical DT-109, Purity: ≥95%, CAS: 14857-82-0, 10 g
SDP
Catalog No. 502944443
Encompass_Preferred
A tripeptide; reduces blood glucose levels in an OGTT in mice; reduces non-fasting blood glucose levels, serum total cholesterol and LDL levels, and adipocyte hypertrophy in Apo-/-- mice at 1 mg/g per day; reduces body fat mass, adipocyte hypertrophy, and hepatomegaly in a mouse model of diet-induced NASH; reverses hepatic steatosis and prevents the progression of fibrosis in a cynomolgus monkey model of NASH induced by a high-fat, high-fructose, and high-cholesterol diet
Catalog No. 50-294-4443 Supplier Cayman Chemical Supplier No. 4121910 G
A tripeptide; reduces blood glucose levels in an OGTT in mice; reduces non-fasting blood glucose levels, serum total cholesterol and LDL levels, and adipocyte hypertrophy in Apo-/-- mice at 1 mg/g per day; reduces body fat mass, adipocyte hypertrophy, and hepatomegaly in a mouse model of diet-induced NASH; reverses hepatic steatosis and prevents the progression of fibrosis in a cynomolgus monkey model of NASH induced by a high-fat, high-fructose, and high-cholesterol diet