Learn More
Cayman Chemical ANTIBODY Dth 500ug
SDP
Catalog No. 502918919
Encompass_Preferred
A PROTAC that drives aptamer-defined protein degradation; composed of DFHBI and thalidomide; induces degradation of mCherry in HEK293T cells at 20 µM; induces degradation of p50, as well as p50 and E2F1, in MCF-7 cells at 20 µM
Catalog No. 50-291-8919 Supplier Cayman Chemical Supplier No. 38925500 UG
A PROTAC that drives aptamer-defined protein degradation; composed of DFHBI and thalidomide; induces degradation of mCherry in HEK293T cells at 20 µM; induces degradation of p50, as well as p50 and E2F1, in MCF-7 cells at 20 µM