Learn More
Cayman Chemical DTUN, 10 mg
SDP
Catalog No. 502947060
Encompass_Preferred
A lipophilic hyponitrite radical initiator; initiates co-autoxidation of STY-BODIPY-embedded egg phosphatidylcholine liposomes at 0.2 mM; has been used in FENIX assays for the development of ferroptosis inhibitors and the study of antioxidants in lipid membranes
Catalog No. 50-294-7060 Supplier Cayman Chemical Supplier No. 3274210 MG
A lipophilic hyponitrite radical initiator; initiates co-autoxidation of STY-BODIPY-embedded egg phosphatidylcholine liposomes at 0.2 mM; has been used in FENIX assays for the development of ferroptosis inhibitors and the study of antioxidants in lipid membranes