Learn More
Cayman Chemical DU-14, Purity: ≥95%, CAS: 186303-55-9, 5 mg
SDP
Catalog No. 502918445
Encompass_Preferred
A steroid sulfatase inhibitor (IC50 = 55.8 nM); inhibits the proliferation of MCF-7 breast cancer cells (IC50 = 38.7 nM); potentiates DHEAS reversal of scopolamine-induced amnesia in the passive avoidance test in rats at 30 mg/kg; prevents scopolamine-induced amnesia in the passive avoidance test in rats at 30 mg/kg per day
Catalog No. 50-291-8445 Supplier Cayman Chemical Supplier No. 396655 MG
A steroid sulfatase inhibitor (IC50 = 55.8 nM); inhibits the proliferation of MCF-7 breast cancer cells (IC50 = 38.7 nM); potentiates DHEAS reversal of scopolamine-induced amnesia in the passive avoidance test in rats at 30 mg/kg; prevents scopolamine-induced amnesia in the passive avoidance test in rats at 30 mg/kg per day