Learn More
Cayman Chemical ANTIBODY DUPA 10mg
SDP
Catalog No. 502982184
Encompass_Preferred
An inhibitor of PSMA (IC50 = 47 nM for the glutamate carboxypeptidase activity); has been used as a ligand to target radiolabels, cytotoxic agents, and antibody-drug conjugates to cancer cells
Catalog No. 50-298-2184 Supplier Cayman Chemical Supplier No. 1950110 MG
An inhibitor of PSMA (IC50 = 47 nM for the glutamate carboxypeptidase activity); has been used as a ligand to target radiolabels, cytotoxic agents, and antibody-drug conjugates to cancer cells