Learn More
Cayman Chemical ANTIBODY eCF309 500ug
SDP
Catalog No. 502981682
Encompass_Preferred
A selective inhibitor of mTOR (IC50 = 15 nM in vitro and in cells)
Catalog No. 50-298-1682 Supplier Cayman Chemical Supplier No. 21118500 UG
A selective inhibitor of mTOR (IC50 = 15 nM in vitro and in cells)