Learn More
Cayman Chemical Eledoisin, Purity: ≥98%, CAS: 69-25-0, 1 mg
SDP
Catalog No. 502937521
Encompass_Preferred
An undecapeptide tachykinin neuropeptide and NK receptor agonist; binds to NK1 (pD2 = 9.5 and 9.21 in isolated dog carotid artery and guinea pig ileum, respectively), NK2 (pD2 = 8.22 and 7.7 in isolated rabbit pulmonary artery and rat duodenum, respectively), and NK3 (pD2 = 7.11 and 7.06 in isolated rat portal vein and hamster urinary bladder, respectively); induces contraction of isolated guinea pig ileum and rabbit jejunum; induces salivation in rats; lowers arterial blood pressure in rabbits and guinea pigs in vivo
Catalog No. 50-293-7521 Supplier Cayman Chemical Supplier No. 288101 MG
An undecapeptide tachykinin neuropeptide and NK receptor agonist; binds to NK1 (pD2 = 9.5 and 9.21 in isolated dog carotid artery and guinea pig ileum, respectively), NK2 (pD2 = 8.22 and 7.7 in isolated rabbit pulmonary artery and rat duodenum, respectively), and NK3 (pD2 = 7.11 and 7.06 in isolated rat portal vein and hamster urinary bladder, respectively); induces contraction of isolated guinea pig ileum and rabbit jejunum; induces salivation in rats; lowers arterial blood pressure in rabbits and guinea pigs in vivo