Learn More
Cayman Chemical Eletriptan, Purity: ≥98%, CAS: 143322-58-1, 50 mg
SDP
Catalog No. 502984494
Encompass_Preferred
An agonist of 5-HT1B and 5-HT1D receptors; induces vasoconstriction of isolated rabbit common carotid artery rings (pD2 = 4.9); preferentially induces constriction of isolated human cerebral over coronary arteries (EC50s = 15.8 and 1,995 nM, respectively)
Catalog No. 50-298-4494 Supplier Cayman Chemical Supplier No. 2004850 MG
An agonist of 5-HT1B and 5-HT1D receptors; induces vasoconstriction of isolated rabbit common carotid artery rings (pD2 = 4.9); preferentially induces constriction of isolated human cerebral over coronary arteries (EC50s = 15.8 and 1,995 nM, respectively)