Learn More
Cayman Chemical Elinogrel, Purity: ≥98%, CAS: 936500-94-6, 1 mg
SDP
Catalog No. 502933370
Encompass_Preferred
A reversible antagonist of P2Y12 (Ki = 23 nM); inhibits platelet aggregation in an hPRP assay (IC50 = 2.81 nM); inhibits thrombosis in a mouse model of vascular injury induced by FeCl3 at 60 mg/kg
Catalog No. 50-293-3370 Supplier Cayman Chemical Supplier No. 259761 MG
A reversible antagonist of P2Y12 (Ki = 23 nM); inhibits platelet aggregation in an hPRP assay (IC50 = 2.81 nM); inhibits thrombosis in a mouse model of vascular injury induced by FeCl3 at 60 mg/kg