Learn More
Cayman Chemical ELR510444, Purity: ≥95%, CAS: 1233948-35-0, 25 mg
SDP
Catalog No. 502980041
Encompass_Preferred
An inhibitor of tubulin polymerization (IC50 = 10 μM); binds to tubulin at the colchicine binding site; induces microtubule depolymerization in A-10 cells (EC50 = 21 nM); inhibits growth in a cancer cell line panel (IC50s = 9-43 nM) via formation of multiple spindles and induction of mitotic arrest; reduces tumor size in an MDA-MB-231 breast cancer mouse xenograft model in a dose-dependent manner; inhibits HIF-1α in RCC4 cells in a concentration-dependent manner
Catalog No. 50-298-0041 Supplier Cayman Chemical Supplier No. 1769625 MG
An inhibitor of tubulin polymerization (IC50 = 10 μM); binds to tubulin at the colchicine binding site; induces microtubule depolymerization in A-10 cells (EC50 = 21 nM); inhibits growth in a cancer cell line panel (IC50s = 9-43 nM) via formation of multiple spindles and induction of mitotic arrest; reduces tumor size in an MDA-MB-231 breast cancer mouse xenograft model in a dose-dependent manner; inhibits HIF-1α in RCC4 cells in a concentration-dependent manner