Learn More
Cayman Chemical EMPO, Purity: ≥95%, CAS: 61856-99-3, 50 mg
SDP
Catalog No. 502983080
Encompass_Preferred
A hydrophilic cyclic nitrone free radical spin trap; EMPO-superoxide adduct that exhibits a t1/2 of 4.8 minutes
Catalog No. 50-298-3080 Supplier Cayman Chemical Supplier No. 2061850 MG
A hydrophilic cyclic nitrone free radical spin trap; EMPO-superoxide adduct that exhibits a t1/2 of 4.8 minutes