Learn More
Cayman Chemical X-376, Purity: ≥98%, CAS: 1365267-27-1, 25 mg
SDP
Catalog No. 502980896
Encompass_Preferred
An ALK and MET inhibitor (IC50s = 0.61 and 0.69 nM, respectively); selectively inhibits the growth of cancer cell lines harboring gain-of-function ALK-fusion proteins (IC50s = 32-150 nM) over those with cell growth driven by other mutant kinases (IC50s = 1,427->3,000 nM); reduces tumor growth in an NCI H3122 lung cancer mouse xenograft model at 50 mg/kg
Catalog No. 50-298-0896 Supplier Cayman Chemical Supplier No. 2134125 MG
An ALK and MET inhibitor (IC50s = 0.61 and 0.69 nM, respectively); selectively inhibits the growth of cancer cell lines harboring gain-of-function ALK-fusion proteins (IC50s = 32-150 nM) over those with cell growth driven by other mutant kinases (IC50s = 1,427->3,000 nM); reduces tumor growth in an NCI H3122 lung cancer mouse xenograft model at 50 mg/kg