Learn More
Cayman Chemical ANTIBODY ERD-308 500ug
SDP
Catalog No. 502946537
Encompass_Preferred
A PROTAC that drives ER degradation; induces degradation of ERα in MCF-7 and T47D cancer cells (DC50s = 0.17 and 0.43 nM, respectively); inhibits the growth of MCF-7 cells (GI50 = 0.77 nM); reduces the levels of mRNA encoding progesterone receptor and GREB1 in MCF-7 cells at 1 to 300 nM
Catalog No. 50-294-6537 Supplier Cayman Chemical Supplier No. 33388500 UG
A PROTAC that drives ER degradation; induces degradation of ERα in MCF-7 and T47D cancer cells (DC50s = 0.17 and 0.43 nM, respectively); inhibits the growth of MCF-7 cells (GI50 = 0.77 nM); reduces the levels of mRNA encoding progesterone receptor and GREB1 in MCF-7 cells at 1 to 300 nM