Learn More
Cayman Chemical Etretinate, Purity: ≥98%, CAS: 54350-48-0, 50 mg
SDP
Catalog No. 502983021
Encompass_Preferred
A second-generation retinoid previously used systemically to manage psoriasis but withdrawn due to teratogenicity; alters cAMP signaling, which may be defective in psoriasis
Catalog No. 50-298-3021 Supplier Cayman Chemical Supplier No. 1987850 MG
A second-generation retinoid previously used systemically to manage psoriasis but withdrawn due to teratogenicity; alters cAMP signaling, which may be defective in psoriasis