Learn More
Cayman Chemical ANTIBODY Exo1 50mg
SDP
Catalog No. 502979385
Encompass_Preferred
A cell-permeable, reversible inhibitor of exocytosis (IC50 = 20 µM); interferes with protein trafficking and secretion mediated by the Golgi apparatus and endoplasmic reticulum by dissociating the GTPase activator ARF1 from Golgi membranes
Catalog No. 50-297-9385 Supplier Cayman Chemical Supplier No. 1845550 MG
A cell-permeable, reversible inhibitor of exocytosis (IC50 = 20 µM); interferes with protein trafficking and secretion mediated by the Golgi apparatus and endoplasmic reticulum by dissociating the GTPase activator ARF1 from Golgi membranes