Learn More
Viro Labs Antibody Ex Enh Susp Cells 10X
Catalog No. NC3728511
Encompass
Size: 250 mLAntibody Expression Enhancer for Suspension Cells (10X)Features:Dramatically boost antibody productivityDramatically increase cell growth and viabilityCompatible with most media formulationsDefined protein-free formulations without any animal-derived components
Catalog No. NC3728511 Supplier Viro Labs Supplier No. ABEH01
Size: 250 mLAntibody Expression Enhancer for Suspension Cells (10X)Features:Dramatically boost antibody productivityDramatically increase cell growth and viabilityCompatible with most media formulationsDefined protein-free formulations without any animal-derived components