Learn More
Viro Labs Antibody Ex Enh Susp Cells 10X
Supplier:  Viro Labs ABEH02
Encompass
Size: 250 mLAntibody Expression Enhancer with Serum for Suspension Cells (10X)FEATURES:Convenient to use.Dramatically boost antibody productivity.Dramatically increase cell growth and viability. Compatible with most media formulations.
Catalog No. NC3728515