Learn More
Cayman Chemical Ezetimibe, Purity: ≥98%, CAS: 163222-33-1, 25 mg
SDP
Catalog No. 502970505
Encompass_Preferred
An inhibitor of NPC1L1 (Ki = 220 nM); reduces cholesterol absorption in hamsters fed a high-cholesterol diet (ED50 = 0.04 mg/kg per day); reduces the levels of helper T cells and memory T cells from the blood of heart transplant recipients at 346 nM
Catalog No. 50-297-0505 Supplier Cayman Chemical Supplier No. 1633125 MG
An inhibitor of NPC1L1 (Ki = 220 nM); reduces cholesterol absorption in hamsters fed a high-cholesterol diet (ED50 = 0.04 mg/kg per day); reduces the levels of helper T cells and memory T cells from the blood of heart transplant recipients at 346 nM