Learn More
Cayman Chemical F2201, Purity: ≥98%, CAS: 1391485-39-4, 10 mg
SDP
Catalog No. 502895090
Encompass_Preferred
F2201 is an analog of AM2201 with a fluorine at the 4 position of the naphthyl group. The physiological and toxicological properties of this compound have not been delineated. This product is intended for research and forensic purposes.
Catalog No. 50-289-5090 Supplier Cayman Chemical Supplier No. 900158010 MG
F2201 is an analog of AM2201 with a fluorine at the 4 position of the naphthyl group. The physiological and toxicological properties of this compound have not been delineated. This product is intended for research and forensic purposes.