Learn More
Cayman Chemical FAB-144, Purity: ≥98%, CAS: 2180935-79-7, 10 mg
SDP
Catalog No. 502971902
Encompass_Preferred
An analog of XLR11 that has an indazole, rather than an indole group; intended for forensic and research applications
Catalog No. 50-297-1902 Supplier Cayman Chemical Supplier No. 1515510 MG
An analog of XLR11 that has an indazole, rather than an indole group; intended for forensic and research applications